AIN-76 vitamin mixture
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ICNA0290545401EA 214 EUR
ICNA0290545401
AIN-76 vitamin mixture
Dierenbenodigdheden Voeding voor dieren
AIN-76 vitamin mixture is a basic component of animal diet for mice and rats colonies in the research lab.
Please see product datasheet for complete diet composition.
Verpakking: In powdered form.