Soy Flour, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0296002405 0296002425
ICNA0296002405EA 53.3 EUR
ICNA0296002405 ICNA0296002425
Soy Flour, MP Biomedicals
Dierenbenodigdheden Voeding voor dieren
Soy flour or soy grits refers to defatted soybeans ground finely.
  • Crude Protein > 48%, Crude Fat > 0.5%, Crude Fibre < 3.5%
  • Used in animal research diet