Vitamin mixture, AIN-93-VX
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ICNA0296040201EA 214 EUR
ICNA0296040201
Vitamin mixture, AIN-93-VX
Dierenbenodigdheden Voeding voor dieren
AIN-93-VX vitamin mixture is a component of AIN-93M and AIN-93G diet.
This vitamin mix is used for the rodents diet revised after numerous nutritional and technical problems were encountered, especially during growth, pregnancy, and lactation.
Opgelet: Not available in pellet form.