pH combination electrodes, Orion™ Economy Series
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
912600 913600 9106BNWP 911600
662-0784EA 515 EUR
662-0784 662-0785 662-1529 662-1454
pH combination electrodes, Orion™ Economy Series
Elektroden pH-elektroden
pH combination electrodes for routine pH measurements.
  • Wick junction, permanently gelled Ag/AgCl electrolyte
  • Unbreakable epoxy body extends beyond the pH sensing glass bulb: Suitable for rough conditions
  • Low-maintenance design requires no fill solution
  • Durability for lab or field use