3-in-1 pH/ATC probes, Orion™ Triode™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9157BN 9157BNMD
662-0787EA 580 EUR
662-0787 662-0986
3-in-1 pH/ATC probes, Orion™ Triode™
Elektroden pH-elektroden
These versatile 3-in-1 pH/ATC probes can be used in either the field or a plant.
  • Type reference: Ag/AgCl
  • Ag/AgCl internal reference system, sealed reference
  • Refillable
Leveringsinformatie: Supplied with one 60 ml bottle of filling solution.