Redox/ORP/temperature electrodes, Orion™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9179BN 9179BNMD 9180BNMD
ORIO9179BNEA 625 EUR
ORIO9179BN 662-1456 662-1457
Redox/ORP/temperature electrodes, Orion™
Elektroden Redox Electrodes
These ORP electrodes combine a platinum redox sensing electrode and a silver/silver chloride reference electrode in one body.
  • Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance
  • Available in epoxy triode low maintenance and epoxy refillable triode construction
  • ORP Triode electrodes have a built-in thermistor for measuring temperature
The 9180BNMD and 9179BNMD ORP/ATC Triode electrodes have a temperature connector that is compatible with the Star Series meters.
The 9180BN and 9179BN ORP/ATC Triode electrode have a temperature connector that is compatible with the A+ Series meters.
Note: Not designed for amperometric or constant current titrations, such as Karl Fischer analysis.