Ion selective electrodes, fluoride, Orion™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9409SC 9609BNWP 9409BN
662-0830EA 1300 EUR
662-0830 662-1539 662-1517
Ion selective electrodes, fluoride, Orion™
Elektroden ISE-elektroden
These fluoride electrodes are designed for measuring free fluoride ions in aqueous solutions quickly, simply, accurately and economically.
  • Stable, drift-free potentials
  • Solid-state
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
Bestelinformatie: The half cell electrodes require a separate reference electrode which can be ordered separately.