Ion selective electrodes, nitrate, Orion™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9307BNWP 9707BNWP
662-1589EA 1590 EUR
662-1589 662-1576
Ion selective electrodes, nitrate, Orion™
Elektroden ISE-elektroden
These nitrate electrodes are designed for measuring nitrate in aqueous solutions quickly, simply, accurately and economically.
  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • Combination electrode 9707BNWP does not require a separate reference electrode, making it convenient to use with small sample volumes
Bestelinformatie: The half cell electrodes require a separate reference electrode which can be ordered separately.