Ion selective electrodes, calcium, Orion™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9320BN 9720BNWP
662-0819EA 2360 EUR
662-0819 662-1579
Ion selective electrodes, calcium, Orion™
Elektroden ISE-elektroden
These calcium electrodes are designed for measuring calcium in aqueous solutions simply, accurately and economically.
  • Built-in Sure-Flow™ reference junction prevents electrode clogging and provides fast and stabile readings
  • Combination electrode does not require a separate reference electrode, making it convenient to use with small sample volumes
Bestelinformatie: The half cell electrode requires a separate reference electrode which can be ordered separately.