Ion selective electrodes, iodide, Orion™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9453BN 9653BNWP 9453SC
662-0845EA 1390 EUR
662-0845 662-1574 ORIO9453SC
Ion selective electrodes, iodide, Orion™
Elektroden ISE-elektroden
These electrodes are designed for measuring iodide ions in quickly, simply, accurately and economically.
  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode does not require a separate reference electrode, making it convenient to use with small sample volumes
Bestelinformatie: The half cell electrodes require a separate reference electrode which can be ordered separately.