Protective guards for conductivity cells and dissolved oxygen probes, Orion™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORIO081045EA 260 EUR
ORIO081045
Protective guards for conductivity cells and dissolved oxygen probes, Orion™
Elektrodehouders
These protective guards provide additional protection and weight for conductivity cells and DO probes.
The plastic guard should be used when when minimal weight and protection are required. The combined guard with stainless steel cap and plastic-sleeve is ideal for use when weight is needed for submersion and standard protection is needed.
For use with:
Conductivity cells: 013005MD, 013010MD, 0130020MD, 013025MD, 013005A, 013010A, and 013005D
Dissolved oxygen probes: 080510MD, 0830005MD, 083010MD, 083025MD, 083060MD, 083005A, 083010A, and 083005D.