Pheos CX2 Safety Spectacles
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9198.285 9198.237 9198.256 9198.257 9198.258 9198.064 9198.275 9198.223
111-2152EA 24.5 EUR
111-2152 111-2148 111-2149 111-2150 111-2151 111-2387 UVEX9198.275 UVEX9198.223
Pheos CX2 Safety Spectacles
Brillen
Modern, sportive designed safety glasses featuring innovative uvex duo component technology for maximum wearer comfort.
  • Comfortable protection due to directly injected forehead and nose pad
  • Optimal protection due to twist temple technology for a perfect and non-slip fit
  • Duo spherical lens provides a perfect field of vision
  • Excellent ventilation ensures a well eye climate
  • A slim temple geometry enables the combination of the spec with other PPE (earmuffs, helmets and caps)
  • CBR65 model: improved contrast; bluelight reduction (ideal for working in the dark); relaxed vision for less fatigue; 65% transmission
Certificaten: EN 166, EN 170