SMART Digest™ ImmunoAffinity (IA) kits
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60110-101 60110-102 60110-103 60110-104 60111-101 60111-102 60111-103 60111-104 60112-101 60112-102 60112-103 60112-104
555-0677EA 0 EUR
555-0677 HYPE60110-102 HYPE60110-103 555-0678 555-0679 HYPE60111-102 HYPE60111-103 555-0680 555-0681 HYPE60112-102 HYPE60112-103 555-0682
SMART Digest™ ImmunoAffinity (IA) kits
Eiwitzuivering Protein Purification Kits
Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Kits are designed to provide fast, simple, highly reproducible and sensitive immunocapture and protein digestion in a single well. These kits remove the challenges often associated with the process of biomarker quantitation, by providing a simple and easy capture-digestion workflow that takes only hours instead of overnight.
  • Significantly faster than traditional protocols
  • Easy to use
  • Highly sensitive and reproducible
  • Compatible with automation
The SMART Digest™ IA Kits are uniquely designed whereby the immunoaffinity reagents (either streptavidin, protein A or protein G) and heat activated thermally stable trypsin are co-immobilised onto a single bead. Following the binding of a capture reagent to the bead, and enrichment of the target, the enzyme is activated at elevated temperatures for accelerated digestion under protein denaturing conditions. The resulting workflow is as easy as enrich, wash and digest. Magnetic and non-magnetic versions of the beads are available.
Leveringsinformatie: Kits are available with magnetic or non magnetic beads, and with or without SOLAµ™ SPE/collection plates.