Gas sampling bags, Tedlar®
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Tedlar®
REST22049EA 427 EUR
REST22049 REST22050 REST22051 REST22052 REST22053 REST22054 REST22055 REST22056 REST22057 REST22058
Gas sampling bags, Tedlar®
Zakken Gaszakken
These sampling bags are a low-cost, whole-air sampling device for high-level VOCs and permanent gases. Made of polyvinyl fluoride (PVF) polymer resin and with PP valve and septum fitting.
  • Lightweight and easy to use
  • Film type can be chosen based on the application
  • Continuous sampling temperature up to 107 °C; short term (1 to 2 hours) temperature up to 176 °C
Several EPA, NIOSH, and OSHA methods exist for bag sampling for a variety of applications: stationary sources emissions; workplace atmospheres; ambient, indoor air quality; and breath analysis. A single eyelet provides handling convenience.