Dulbecco's phosphate buffered saline (DPBS), cell culture, MP Biomedicals
Leverancier: MP Biomedicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
091860449
091860454
091860454X6
ICNA091860449EA
17.1
EUR
ICNA091860449
ICNA091860454
ICNA091860454X6
Dulbecco's phosphate buffered saline (DPBS), cell culture, MP Biomedicals
Buffers
Buffers voor weefselkweek
Phosphate buffered salts, Dulbecco's formula (DPBS) is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells. DPBS is used to to irrigate, wash, and dilute mammalian cells.
- Clear, colourless solution format
- pH: 7,0 to 7,6
- Stored at 15 to 30 °C
Phosphate buffering maintains the pH in the physiological range. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells.