Human plasma, defibrinated, delipidated
Leverancier: SEQENS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30010-100ML 30010-500ML 30010-1L
30010-100MLEA 180 EUR
30010-100ML 30010-500ML 30010-1L
Human plasma, defibrinated, delipidated
Plasma
SEQENS IVD manufactures Defibrinated Delipidated Human Plasma. It is dialysed and electrolytes balanced.
  • Dry extract: 65 to 85 g/L
  • Total proteins: 55 to 75 g/L
  • Fibrinogen: Absent
  • Cholesterol: <0,04 g/L
  • Triglycerides: <0,05 g/L
  • Filtered through a 0,2 µm membrane
  • Shelf life: 2 years
For this product, source material has been tested and found negative or non-reactive (FDA approved and/or CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.
Complete donor traceability on file.
Opgelet: Only for research or manufacture of in vitro diagnostic reagents. Not for therapeutic use. This product of human origin should be considered as potentially infectious and handled accordingly.