Human processed serum (HPS), Vitamin D depleted
Leverancier: SEQENS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
33000D-100ML 33000D-500ML 33000D-1L
33000D-100MLEA 206 EUR
33000D-100ML 33000D-500ML 33000D-1L
Human processed serum (HPS), Vitamin D depleted
Serum
SEQENS IVD manufactures Vitamin D Depleted Human Processed Serum.
  • Dry extract: 65 to 85 g/L
  • Total proteins: 55 to 75 g/L
  • Vitamin 25-OH D3: <4 ng/ml
  • Filtered through a 0,2 µm membrane
  • Shelf life: 2 years
For this product, source material has been tested and found negative or non-reactive (FDA approved and/or CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.
Complete donor traceability on file.
Opgelet: Only for research or manufacture of in vitro diagnostic reagents. NotMore Product Information