Packing material for columns, Res-Sil
Leverancier: Restek
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25028
25030
25042
25044
25080
25081
25400
25401
REST25028EA
80.4
EUR
REST25028
REST25030
REST25042
REST25044
REST25080
REST25081
REST25400
REST25401
Packing material for columns, Res-Sil
Sorbenten voor chromatografie
Sorbenten voor GC
These bonded silica packings with n-octane or cyanopropyl (OPN) functional groups yield faster separations of C1 to C4 hydrocarbons, higher thermal stability, shorter conditioning times, and longer lifetimes.
- Unique separation of saturated and unsaturated hydrocarbons
- Innovative bonding chemistry for batch-to-batch reproducibility
Each production batch of Res-Sil C packing is tested with a complex hydrocarbon mixture to meet demanding retention time and retention index specifications.