VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85518.260EA
25.3
EUR
85518.260
85511.260
85515.260
85516.260
85517.260
85506.260
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Buffers
pH-buffers
- Certified Reference Materials (CRMs) according to ISO 17034
- Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents
- True analytical value on the label to an accuracy of ±0,02 pH units
- Traceable to SRM from NIST
- Batch number and packing date for traceability
Certificates of Analysis available at website
Certificaten: Produced and certified in accordance with ISO 17034 accredited by BELAC under the certification number 542-RM. All information regarding the use of this CRM can be found on the Certificate of Analysis.