VWR® ARISTAR®, Aluminium Standard Solutions for ICP-MS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85548.180EA
149
EUR
85548.180
457202A
VWR® ARISTAR®, Aluminium Standard Solutions for ICP-MS
Standaarden
Kationische standaarden
Aluminium standaarden
- Produced from high purity acids, water ASTM I 18 Megaohms
- Solution assayed by titration
- Final concentration verified against reference material from NIST
- Total maximum uncertainty: 0,5% (typical)
- Delivered with complete Certificate of Analysis
1000 ppm equivalent to 1000 mg/L
Certificaten: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.