Conductivity standards, KCl, referenced at 25 °C
Leverancier: REAGECON
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CSKC10
CSKC100
CSKC1000
CSKC100M
CSKC10M
CSKC12880
CSKC150M
CSKC20
CSKC200
CSKC200M
CSKC20M
CSKC300M
CSKC3M
CSKC5
CSKC50
CSKC500
CSKC50M
CSKC5M
CSKC84
CSKCL
CSKCS
CSKC136
CSKC350M
CSKC500M
CSKC13
CSKC2M
CSKC80M
CSKC450M
CSKC238
CSKC300
CSKC400
CSKC111800
CSKC2500
CSKC25
CSKC185
CSKC250
CSKC30M
CSKC400M
CSKC60M
CSKC40M
CSKC1025
REGECSKC10EA
282
EUR
REGECSKC10
REGECSKC100
REGECSKC1000
REGECSKC100M
REGECSKC10M
REGECSKC12880
REGECSKC150M
REGECSKC20
REGECSKC200
REGECSKC200M
REGECSKC20M
REGECSKC300M
REGECSKC3M
REGECSKC5
REGECSKC50
REGECSKC500
REGECSKC50M
REGECSKC5M
REGECSKC84
REGECSKCL
REGECSKCS
REGECSKC136
REGECSKC350M
REGECSKC500M
REGECSKC13
REGECSKC2M
REGECSKC80M
REGECSKC450M
REGECSKC238
REGECSKC300
REGECSKC400
REGECSKC111800
REGECSKC2500
REGECSKC25
REGECSKC185
REGECSKC250
REGECSKC30M
REGECSKC400M
REGECSKC60M
REGECSKC40M
REGECSKC1025
Conductivity standards, KCl, referenced at 25 °C
Standaarden
Geleidbaarheidsstandaarden
These conductivity standards are aqueous and so avoid the problems of matrix errors and high temperature dependency of nonaqueous conductivity standards.
- With an accuracy of ±1,0% (except 1,3 µS)
- Temperature dependence data, lot no. and expiry date are printed on product labels
These solutions standardised at 25 °C are directly traceable to N.I.S.T standard reference materials and are tested using an ISO 17025 accredited method.