Fluorescence cuvettes
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6610010400 6610013800 6610015400 6610016000 6610016100 6610016200 6610016300 6610016400 6610019600 6610019700 6610019800 6610023900 6610024000 6610024100 6618000100 6610000800 6610001100 6610001800 6610008800
VARI6610010400EA 520 EUR
VARI6610010400 VARI6610013800 VARI6610015400 VARI6610016000 VARI6610016100 VARI6610016200 VARI6610016300 VARI6610016400 VARI6610019600 VARI6610019700 VARI6610019800 VARI6610023900 VARI6610024000 VARI6610024100 VARI6618000100 VARI6610000800 VARI6610001100 VARI6610001800 VARI6610008800
Fluorescence cuvettes
Cuvetten Cuvetten voor spectrofotometers
Fluorescence cuvettes designed for UV/visible analysis in routine applications of molecular spectroscopy.
  • Longer pathlength cells for extra sensitivity in low concentration samples
Suitable for low concentration or highly corrosive samples.
Longer pathlength (20 - 50 mm) cells are suitable for extra sensitivity in low concentration samples.
Bestelinformatie: 10 mm pathlength models are available in matched pairs.