VWR® ARISTAR®, Cadmium Standard Solutions for ICP-MS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456682REA
164
EUR
456682R
85556.180
VWR® ARISTAR®, Cadmium Standard Solutions for ICP-MS
Standaarden
Kationische standaarden
Cadmium standaarden
- Produced from high purity acids, water ASTM I 18 Megaohms
- Solution assayed by titration
- Final concentration verified against reference material from NIST
- Total maximum uncertainty: 0,5% (typical)
- Delivered with complete certificate of analysis
Certificaten: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities