VWR® ARISTAR®, Phosphorus Standard Solutions for ICP-MS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456842PEA
167
EUR
456842P
457112P
85585.180
VWR® ARISTAR®, Phosphorus Standard Solutions for ICP-MS
Standaarden
Kationische standaarden
Fosfor standaarden
- Produced from high purity acids, water ASTM I 18 Megaohms
- Solution assayed by titration
- Final concentration verified against reference material from NIST
- Total maximum uncertainty: 0.5% (typical)
- Delivered with complete certificate of analysis
Certificaten: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities