VWR® ARISTAR®, Lead Standard Solutions for ICP-MS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456772SEA 174 EUR
456772S 85586.180
VWR® ARISTAR®, Lead Standard Solutions for ICP-MS
Standaarden Kationische standaarden Lood standaarden
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0.5% (typical)
  • Delivered with complete certificate of analysis
Certificaten: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities
Order Now
Specificaties
Environmentally Preferable