VWR®, Zinc standard solutions for ICP-MS, ARISTAR®
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456992FEA 174 EUR
456992F 457162C 85648.180
VWR®, Zinc standard solutions for ICP-MS, ARISTAR®
Standaarden Kationische standaarden Zink standaarden
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis
Certificaten: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities