Anti-BAX Mouse Monoclonal Antibody (CF640R) [clone: 2D2]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC400004-500 BNC400004-100
BNC400004-500EA 560 EUR
BNC400004-500 BNC400004-100
Anti-BAX Mouse Monoclonal Antibody (CF640R) [clone: 2D2]
Antilichamen
Recognizes a protein of 21 kDa, identified as the Bax protein. This MAb is highly specific to Bax and shows no cross-reaction with Bcl-2 or Bcl-X protein. Bcl-2 blocks cell death following a variety of stimuli. Bax has extensive amino acid homology with Bcl-2 and it homodimerizes and forms heterodimers with Bcl-2. Overexpression of Bax accelerates apoptotic death induced by cytokine deprivation in an IL-3 dependent cell line, and Bax also counters the death repressor activity of Bcl-2.
Type: Primary
Antigen: BAX
Clonality: Monoclonal
Clone: 2D2
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey