Anti-CDKN1A Mouse Monoclonal Antibody (Purified) [clone: WA-1]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNUB0043-500 BNUB0043-100
BNUB0043-500EA 511 EUR
BNUB0043-500 BNUB0043-100
Anti-CDKN1A Mouse Monoclonal Antibody (Purified) [clone: WA-1]
Antilichamen
This MAb recognizes a 21 kDa protein, identified as the p21WAF1 tumor suppressor protein. This MAb is highly specific to p21 and shows no cross-reaction with other closely related mitotic inhibitors. p21WAF1 is a specific inhibitor of cdk s and a tumor suppressor involved in the pathogenesis of a variety of malignancies. The expression of this gene acts as an inhibitor of the cell cycle during G1 phase and is tightly controlled by the tumor suppressor protein p53. Its expression is induced by the wild type, but not mutant, p53 suppressor protein. Normal cells generally display a ratherMore Product Information
Order Now

Specificaties

antigeen symbool CDKN1A
Environmentally Preferable
Type antilichaam Primair,Cyclin-dependent kinase inhibitor 1A
Antigeen synoniemen p21|WAF1
Type antilichaam Primair,Cyclin-dependent kinase inhibitor 1A
Clonality Monoklonaal
Conjugatie Onconjugeerd
Clone Clone WA-1
Reactivity Monkey,Chimpanzee,Human,Rat,Mouse
Host Mouse
Gen-ID 370771
Isotype IgG1 kappa
ImmunoChemie Yes
ImmunoFluorescentie Yes
Concentratie 0,2 mg/ml
Opslagbuffer PBS/0,05% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR