Anti-CD74 Mouse Monoclonal Antibody (CF647) [clone: LN-2]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC470056-500 BNC470056-100
BNC470056-500EA 560 EUR
BNC470056-500 BNC470056-100
Anti-CD74 Mouse Monoclonal Antibody (CF647) [clone: LN-2]
Antilichamen
This antibody recognizes a protein of ~35 kDa, identified as CD74 (Workshop IV). CD74 is a type II transmembrane protein which binds to the peptide binding groove of newly synthesized MHC class II alpha/beta heterodimers and prevents their premature association with endogenous polypeptides. CD74 is expressed primarily by antigen presenting cells, such as B-lymphocytes (from before the pre-B cell stage to before the plasma cell stage), macrophages, and monocytes, and many epithelial cells. Anti-CD74 stains predominantly germinal center lymphocytes and B-cell lymphomas, but rarely T-cell lymphomas. Anti-CD74 has been shown to be useful in differentiating atypical fibroxanthoma (-) from malignant fibrous histiocytoma ( ).
Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: LN-2
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Human, Mouse