Anti-Granulocyte Marker Mouse Monoclonal Antibody (CF488A) [clone: BM-1]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC880061-500 BNC880061-100
BTIUBNC880061-500EA 560 EUR
BTIUBNC880061-500 BTIUBNC880061-100
Anti-Granulocyte Marker Mouse Monoclonal Antibody (CF488A) [clone: BM-1]
Antilichamen
Recognizes 183 kDa protein with DNA-binding characteristics, which is identified as a myeloid specific antigen. It reacts with myeloid precursor cells and granulocytes in bone marrow. Its antigen appears to be restricted to M2 and M3 acute myelogenous leukemia (AML) subtypes. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. This MAb reacts with early precursor and mature forms of human myeloid cells. It is useful in the identification of myelogenous leukemias, distinguishing granulocytic sarcomas from lymphoid malignancies and also in the study of differentiation and transformation of human myeloid cells. The biological function of this antigen is not clear, although it has been proposed that it may play a role in the differentiation of myeloid cells.
Type: Primary
Antigen: Granulocyte Marker
Clonality: Monoclonal
Clone: BM-1
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human