Anti-UCHL1 Mouse Monoclonal Antibody (CF488A) [clone: 13C4]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC880147-500 BNC880147-100
BNC880147-500EA 560 EUR
BNC880147-500 BNC880147-100
Anti-UCHL1 Mouse Monoclonal Antibody (CF488A) [clone: 13C4]
Antilichamen
This MAb reacts with a protein of 20-30 kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1). Initially, PGP9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP9.5More Product Information
Order Now

Specificaties

antigeen symbool UCHL1
Type antilichaam Primair,Ubiquitin Carboxyl-Terminal Esterase L1
Environmentally Preferable
Type antilichaam Primair,Ubiquitin Carboxyl-Terminal Esterase L1
Clonality Monoklonaal
Conjugatie CF® 488A
Clone Clone 13C4
Reactivity Zebrafish,Pig,Rabbit,Sheep,Human,Bovine,Guinea pig,Dog,Rat,Mouse
Host Mouse
Gen-ID 518731
Isotype IgG2a kappa
Western blot Yes
ImmunoFluorescentie Yes
Concentratie 0,1 mg/ml
Opslagbuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR