Anti-VCAM1 Mouse Monoclonal Antibody (CF594) [clone: 1.4C3]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC940149-500 BNC940149-100
BNC940149-500EA 560 EUR
BNC940149-500 BNC940149-100
Anti-VCAM1 Mouse Monoclonal Antibody (CF594) [clone: 1.4C3]
Antilichamen
Recognizes a protein of 110 kDa, identified as CD106 (also known as vascular cell adhesion molecule-1 (VCAM-1) and INCAM-100). CD106 is a member of the Ig superfamily of adhesion molecules and is expressed at high levels on cytokine stimulated vascular endothelial cells, and at minimal levels on un-stimulated endothelial cells. It is also present on follicular and inter-follicular dendritic cells of lymph nodes, myoblasts, and some macrophages. CD106 serves as a ligand for leukocyte integrin (VLA-4 or CD49d/CD29) and mediates cell adhesion of leukocytes to activated endothelium. It plays aMore Product Information