Anti-CD74 Mouse Monoclonal Antibody (CF568) [clone: BU45]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC680189-500 BNC680189-100
BNC680189-500EA 560 EUR
BNC680189-500 BNC680189-100
Anti-CD74 Mouse Monoclonal Antibody (CF568) [clone: BU45]
Antilichamen
Recognizes proteins of 33, 35 and 41 kDa, which are identified as various isoforms of CD74. Its epitope is localized in the extracellular domain of CD74. CD74 is a type II transmembrane protein which binds to the peptide binding groove of newly synthesized MHC class II alpha/beta heterodimers and prevents their premature association with endogenous polypeptides. The CD74 molecule plays a critical role in the presentation of peptides, by the MHC class II antigens, to CD4 positive lymphocytes. CD74 is expressed on MHC class II positive cells including B cells, a subset of activated T cells,More Product Information