Anti-IGF1 Mouse Monoclonal Antibody (CF405S) [clone: M23]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC040220-500 BNC040220-100
BTIUBNC040220-500EA 560 EUR
BTIUBNC040220-500 BTIUBNC040220-100
Anti-IGF1 Mouse Monoclonal Antibody (CF405S) [clone: M23]
Antilichamen
This antibody is specific to Insulin-like Growth Factor (IGF-1) and shows minimal cross-reaction with IGF-11, Proinsulin, MSF, and Insulin. IGF-1 is a polypeptide growth factor with two isoforms that are produced by alternative splicing. Isoform 1 is also known as IGF-IB while isoform 2 is known as IGF-IA. IGF-1 stimulates the proliferation of a wide range of cell types including muscle, bone and cartilage tissue. It functions as an autocrine regulator of growth. Activation of IGF system has emerged as a key factor for tumor progression and resistance to apoptosis in many cancers like those of breast, thyroid and colon.
Type: Primary
Antigen: IGF1
Clonality: Monoclonal
Clone: M23
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rabbit, Rat