Anti-FN1 Mouse Monoclonal Antibody (Biotin) [clone: HFN7.1]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNCB0270-500 BNCB0270-100
BNCB0270-500EA 560 EUR
BNCB0270-500 BNCB0270-100
Anti-FN1 Mouse Monoclonal Antibody (Biotin) [clone: HFN7.1]
Antilichamen
Fibronectin is a dimeric glycoprotein of 440 kDa, which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing. This MAb is directed against the peptide core and reacts with both the plasma and cellular forms of fibronectin. It blocks the fibronectin-medicated cell attachment not by disrupting the collagen-fibronectin interaction, but by interfering with the attachment of fibronectin to its receptor on the cell surface.

Type: Primary
Antigen: FN1
Clonality: Monoclonal
Clone: HFN7.1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR