Anti-NKX2-2 Mouse Monoclonal Antibody (CF640R) [clone: NX2/294]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC400294-500 BNC400294-100
BNC400294-500EA 560 EUR
BNC400294-500 BNC400294-100
Anti-NKX2-2 Mouse Monoclonal Antibody (CF640R) [clone: NX2/294]
Antilichamen
Expression of NKX2.2 has been found in neuroendocrine tumors of the gut, making it a potential marker for the study of gastrointestinal neuroendocrine tumors. More recently, NKX2.2 protein was identified as a target of EWS-FLI-1, the fusion protein specific to Ewing sarcoma, and was shown to be differentially upregulated in Ewing sarcoma on the basis of array-based gene expression analysis. It acts as a valuable marker for Ewing sarcoma, with a sensitivity of 93% and a specificity of 89%, and aids in the differential diagnosis of small round cell tumors.

Type: Primary
Antigen: NKX2-2
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR