Anti-Histiocytoma Marker Mouse Monoclonal Antibody (CF647) [clone: D11]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC470348-500 BNC470348-100
BNC470348-500EA 560 EUR
BNC470348-500 BNC470348-100
Anti-Histiocytoma Marker Mouse Monoclonal Antibody (CF647) [clone: D11]
Antilichamen
In Western blotting, this antibody detects an antigen of 125 kDa in human liver and 135 kDa in tumors of histiocytic origin. Comparative study of this MAb and a standard CD68 MAb showed that their antigens are different. Its antigen in all macrophage types studied is located on the plasma membrane and within cytoplasmic structures including lysosomes. This MAb shows a restricted reactivity to cells of the monocyte/macrophage system. It specifically reacts with blood monocytes and stains resident macrophages in a wide variety of human tissues. This MAb does not stain antigen-presenting cells, e.g., Langerhans cells. Reportedly, its reactivity is restricted to histiocytes and macrophages.
Type: Primary
Antigen: Histiocytoma Marker
Clonality: Monoclonal
Clone: D11
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human