Anti-IGHM Mouse Monoclonal Antibody (Purified) [clone: IM373]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNUB0373-500 BNUB0373-100
BNUB0373-500EA 511 EUR
BNUB0373-500 BNUB0373-100
Anti-IGHM Mouse Monoclonal Antibody (Purified) [clone: IM373]
Antilichamen
Recognizes a protein of 75 kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Type: Primary
Antigen: IGHM
More Product Information