Anti-SLC3A2 Mouse Monoclonal Antibody (Biotin) [clone: UM7F8]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNCB0425-500 BNCB0425-100
BNCB0425-500EA 560 EUR
BNCB0425-500 BNCB0425-100
Anti-SLC3A2 Mouse Monoclonal Antibody (Biotin) [clone: UM7F8]
Antilichamen
CD98 exits as a heterodimer containing a disulphide-linked glycosylated heavy chain and a non-glycosylated light chain. It is a member of the solute carrier family and encodes a cell surface, transmembrane protein. The protein exists as the heavy chain of a heterodimer, covalently bound through disulfide bonds to one of several possible light chains. The encoded transporter plays a role in regulation of intracellular calcium levels and transports L-type amino acids. Alternatively spliced transcript variants, encoding different isoforms, have been characterized.
Type: Primary
Antigen: SLC3A2
Clonality: Monoclonal
Clone: UM7F8
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human