Anti-CD6 Mouse Monoclonal Antibody (CF640R) [clone: 3F7B5]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC400428-500 BNC400428-100
BNC400428-500EA 560 EUR
BNC400428-500 BNC400428-100
Anti-CD6 Mouse Monoclonal Antibody (CF640R) [clone: 3F7B5]
Antilichamen
CD6 is a type I transmembrane glycoprotein that contains a 24-amino acid signal sequence, three extracellular scavenger receptor cysteine-rich (SRCR) domains, a membrane-spanning domain and a 44-amino acid cytoplasmic domain. The CD6 glycoprotein is tyrosine phosphorylated during TCR-mediated T cell activation. CD6 shows significant homology to CD5. CD6 is present on mature thymocytes, peripheral T cells and a subset of B cells. Antibodies to CD6 are used to deplete T cells from bone marrow transplants to prevent graft versus host disease.
Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: 3F7B5
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human