Anti-CD19 Mouse Monoclonal Antibody (CF640R) [clone: CVID3/429]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC400429-500 BNC400429-100
BNC400429-500EA 560 EUR
BNC400429-500 BNC400429-100
Anti-CD19 Mouse Monoclonal Antibody (CF640R) [clone: CVID3/429]
Antilichamen
CD19 is a transmembrane glycoprotein that contains two extracellular immunoglobulin-like domains. CD19 is present in both benign and malignant B-cells and is considered to be the most reliable surface marker of this lineage over a wide range of maturational stages. In normal lymphoid tissue, CD19 is observed in germinal centers, in mantle zone cells, and in scattered cells of the inter-follicular areas. Anti-CD19 exhibits an overall immunoreactivity pattern similar to those of the antibodies against CD20 and CD22. However, in contrast to CD20, expression of CD19 is continuous throughout B-More Product Information