Anti-ODC1 Mouse Monoclonal Antibody (CF647) [clone: ODC1/487]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC470487-500 BNC470487-100
BNC470487-500EA 560 EUR
BNC470487-500 BNC470487-100
Anti-ODC1 Mouse Monoclonal Antibody (CF647) [clone: ODC1/487]
Antilichamen
Recognizes a 53 kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those inMore Product Information