Restriction enzymes, NotI, FastDigest™, fermentas
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FD0594
FD0595
FD0594EA
89
EUR
FD0594
FD0595
Restriction enzymes, NotI, FastDigest™, fermentas
Reagentia voor nucleïnezuren
Restriction Enzymes
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
- 100% activity of all FastDigest™ enzymes in one universal reaction buffer
- Designed to complete digestion in 5 to 15 minutes
- Direct loading on gels with FastDigest™ Green buffer
- 100% buffer compatibility with downstream applications