Anti-PGR Mouse Monoclonal Antibody (CF405S) [clone: PR500]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC040500-500 BNC040500-100
BNC040500-500EA 560 EUR
BNC040500-500 BNC040500-100
Anti-PGR Mouse Monoclonal Antibody (CF405S) [clone: PR500]
Antilichamen
This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81 kDa) and PR-B (116 kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone.
Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: PR500
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human