Anti-MYC Mouse Monoclonal Antibody (Purified) [clone: 9E10.]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNUB0596-500 BNUB0596-100
BNUB0596-500EA 511 EUR
BNUB0596-500 BNUB0596-100
Anti-MYC Mouse Monoclonal Antibody (Purified) [clone: 9E10.]
Antilichamen
This antibody recognizes a transcription factor of 64-67 kDa, identified as c-myc. Its epitope spans between aa 410-419 (EQKLISEEDL) which is a specific portion of an alpha helical region of human c-myc protein. This MAb shows no cross-reaction with v-myc. c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors. Over-expression of c-myc protein occurs frequently in luminal cells of prostate intraepithelial neoplasia as well as in most primary carcinomas and metastatic disease.
Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone: 9E10.
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human