Anti-KDR Mouse Monoclonal Antibody (CF640R) [clone: KDR657]

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC400657-500 BNC400657-100
BNC400657-500EA 560 EUR
BNC400657-500 BNC400657-100
Anti-KDR Mouse Monoclonal Antibody (CF640R) [clone: KDR657]
Antilichamen
CD309, also known as VEGF-R2, KDR3, and Flk-1 (mouse), is a type I transmembrane glycoprotein. It is a member of the CSF-1/PDGF receptor family of type III tyrosine kinase receptors. Human VEGF-R2 is mainly expressed by endothelial cells, embryonic tissues, and megakaryocytes. It plays an important role in the regulation of angiogenesis, vasculogenesis, and vascular permeability. The ligands of VEGF-R2 include VEGF-A, VEGF-C, VEGF-D, and VEGF splice isoforms. Ligation of VEGF-R2 with its ligands results in the receptor dimerization and auto-phosphorylation, stimulating endothelial cell proliferation and migration.

Type: Primary
Antigen: KDR
Clonality: Monoclonal
Clone: KDR657
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR