Anti-KDR Mouse Monoclonal Antibody (CF647) [clone: KDR657]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC470657-500 BNC470657-100
BNC470657-500EA 560 EUR
BNC470657-500 BNC470657-100
Anti-KDR Mouse Monoclonal Antibody (CF647) [clone: KDR657]
Antilichamen
CD309, also known as VEGF-R2, KDR3, and Flk-1 (mouse), is a type I transmembrane glycoprotein. It is a member of the CSF-1/PDGF receptor family of type III tyrosine kinase receptors. Human VEGF-R2 is mainly expressed by endothelial cells, embryonic tissues, and megakaryocytes. It plays an important role in the regulation of angiogenesis, vasculogenesis, and vascular permeability. The ligands of VEGF-R2 include VEGF-A, VEGF-C, VEGF-D, and VEGF splice isoforms. Ligation of VEGF-R2 with its ligands results in the receptor dimerization and auto-phosphorylation, stimulating endothelial cell proliferation and migration.
Type: Primary
Antigen: KDR
Clonality: Monoclonal
Clone: KDR657
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human