Anti-KRT10 Mouse Monoclonal Antibody (Purified) [clone: LH2]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNUB0674-500 BNUB0674-100
BNUB0674-500EA 511 EUR
BNUB0674-500 BNUB0674-100
Anti-KRT10 Mouse Monoclonal Antibody (Purified) [clone: LH2]
Antilichamen
This MAb recognizes a protein of 56.5 kDa, identified as cytokeratin 10 (CK10). CK10 is expressed in all suprabasal layers of the epidermis. In the epidermis, expression of CK10 strictly parallels the extent of differentiation; it is absent in the basal layer, appears in the first suprabasal layers and increases in concentration towards the granular layer. However, CK10 is rarely detected in early stages of vulvar squamous carcinomas (tumors less than 2 cm, clinical stage I) regardless of the tumor grade. In larger and more advanced tumors (greater than 2 cm, clinical stages II and III), CK10More Product Information