Anti-Mitochondrial Marker Mouse Monoclonal Antibody (Biotin) [clone: MTC719]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNCB0719-500 BNCB0719-100
BNCB0719-500EA 560 EUR
BNCB0719-500 BNCB0719-100
Anti-Mitochondrial Marker Mouse Monoclonal Antibody (Biotin) [clone: MTC719]
Antilichamen
This MAb recognizes a 60 kDa antigen associated with the mitochondria in cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the mitochondria in cells from a wide variety of animals, but not insects and bacteria. It can be used to stain the mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-like pattern in normal and malignant cells and may be used to stain mitochondria of cells in fixed or frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations.
Type: Primary
Antigen: Mitochondrial Marker
Clonality: Monoclonal
Clone: MTC719
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human