Anti-CD74 Mouse Monoclonal Antibody (CF647) [clone: CLIP/813]
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNC470813-500 BNC470813-100
BNC470813-500EA 560 EUR
BNC470813-500 BNC470813-100
Anti-CD74 Mouse Monoclonal Antibody (CF647) [clone: CLIP/813]
Antilichamen
This antibody recognizes a protein of ~35 kDa, identified as CD74. It is a type II transmembrane protein which binds to the peptide binding groove of newly synthesized MHC class II alpha/beta heterodimers and prevents their premature association with endogenous polypeptides. CD74 is expressed primarily by antigen presenting cells, such as B-lymphocytes (from before the pre-B cell stage to before the plasma cell stage), macrophages, and monocytes, and many epithelial cells. Anti-CD74 stains predominantly germinal center lymphocytes and B-cell lymphomas, but rarely T-cell lymphomas. Anti-CD74More Product Information